Learn More
Medchemexpress LLC 2-Bromo-4'-hydroxyacetophenone | 2491-38-5 | 215.05 | 100 G
SDP
Catalog No. 5000449771
Encompass_Preferred
2-Bromo-4'-hydroxyacetophenone is a PTP1B inhibitor with a Ki value of 42 μM. It also serves as an intermediate and can be used in the synthesis of LXRα/β antagonists. Additionally, it can be utilized in research on hematological diseases.
  • PTP1B inhibitor
  • Intermediate for synthesis of LXRα/β antagonists
  • Used in research on hematological diseases
  • Unstable in solutions, fresh preparation recommended
Catalog No. 50-004-49771 Supplier Medchemexpress LLC Supplier No. HYW002314100G
Add to Cart
Add to Cart
2-Bromo-4'-hydroxyacetophenone is a PTP1B inhibitor with a Ki value of 42 μM. It also serves as an intermediate and can be used in the synthesis of LXRα/β antagonists. Additionally, it can be utilized in research on hematological diseases.
  • PTP1B inhibitor
  • Intermediate for synthesis of LXRα/β antagonists
  • Used in research on hematological diseases
  • Unstable in solutions, fresh preparation recommended