Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromo-4-methylaniline

Catalog No. 12408725G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-408-725G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-408-725G Supplier Sigma Aldrich Supplier No. 12408725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Bromo-4-methylaniline

Specifications

Melting Point 14°C to 16°C (lit.)
CAS 583-68-6
Boiling Point 240°C (lit.)
Molecular Formula C7H8BrN
Linear Formula BrC6H3(CH3)NH2
MDL Number MFCD00007635
Synonym 2-Bromo-p-toluidine; 4-Amino-3-bromotoluene
Molecular Weight (g/mol) 186.05
Percent Purity 98%
Density 1.5 g/mL (at 25°C (literature))
Quantity 100 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.