Learn More
Medchemexpress LLC 2-bromo-4-methylphenol | 6627-55-0 | MFCD00002151 | 99.3% | 187.03 g/mol | C7H7BrO | 100 G
SDP
Catalog No. 5000302261
Encompass_Preferred
2-Bromo-4-methylphenol (CAS 6627-55-0) is a brominated aromatic phenol used as a biochemical reagent and synthetic intermediate in organic and life-science research. Supplied as a colorless to light-yellow liquid, it is intended for laboratory applications requiring a brominated phenolic building block.
  • High purity (99.25%).
  • Molecular formula C7H7BrO; molecular weight 187.03 g/mol.
  • Characterized with COA and SDS available for verification.
  • Suitable as a building block in organic synthesis and reagent for assay development.
  • Available in multiple pack sizes for laboratory use.
Catalog No. 50-003-02261 Supplier Medchemexpress LLC Supplier No. HYW016153100G
2-Bromo-4-methylphenol (CAS 6627-55-0) is a brominated aromatic phenol used as a biochemical reagent and synthetic intermediate in organic and life-science research. Supplied as a colorless to light-yellow liquid, it is intended for laboratory applications requiring a brominated phenolic building block.
  • High purity (99.25%).
  • Molecular formula C7H7BrO; molecular weight 187.03 g/mol.
  • Characterized with COA and SDS available for verification.
  • Suitable as a building block in organic synthesis and reagent for assay development.
  • Available in multiple pack sizes for laboratory use.