Learn More
Medchemexpress LLC 2-bromo-4-methylphenol | 6627-55-0 | MFCD00002151 | 99.3% | 187.03 g/mol | C7H7BrO | 25 G
SDP
Catalog No. 5000304153
Encompass_Preferred
2-Bromo-4-methylphenol is a brominated phenolic reagent used in biochemical research and organic synthesis. It is a high-purity solid (C7H7BrO, 187.03 g/mol) suitable for assay development, small-molecule synthesis, and use as a synthetic building block. The compound is soluble in DMSO and has recommended storage conditions to maintain stability and shelf life.
  • High purity (~99.3%) suitable for analytical and synthetic applications.
  • Soluble in DMSO at 100 mg/mL for concentrated stock solutions.
  • In vivo formulation protocols available for dosing at ≥5 mg/mL.
  • Stable when stored at -20°C in pure form, with extended shelf life under recommended conditions.
  • Useful as a building block for phenolic derivatization and halogenated intermediate synthesis.
Catalog No. 50-003-04153 Supplier Medchemexpress LLC Supplier No. 25GHYW016153
2-Bromo-4-methylphenol is a brominated phenolic reagent used in biochemical research and organic synthesis. It is a high-purity solid (C7H7BrO, 187.03 g/mol) suitable for assay development, small-molecule synthesis, and use as a synthetic building block. The compound is soluble in DMSO and has recommended storage conditions to maintain stability and shelf life.
  • High purity (~99.3%) suitable for analytical and synthetic applications.
  • Soluble in DMSO at 100 mg/mL for concentrated stock solutions.
  • In vivo formulation protocols available for dosing at ≥5 mg/mL.
  • Stable when stored at -20°C in pure form, with extended shelf life under recommended conditions.
  • Useful as a building block for phenolic derivatization and halogenated intermediate synthesis.