Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%

Catalog No. AAH64105MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64105MD 250 mg
AAH6410503 1 g
AAH6410506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH64105MD Supplier Thermo Scientific Chemicals Supplier No. H64105MD

CAS: 103962-10-3 | C9H6BrF3O2 | 283.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103962-10-3
Molecular Formula C9H6BrF3O2
Molecular Weight (g/mol) 283.044
MDL Number MFCD00052333
InChI Key AOAGGWLQIILIIV-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl
PubChem CID 2778691
IUPAC Name 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F

Specifications

Melting Point 48°C to 55°C
Color Yellow
Flash Point >110°C (230°F)
Quantity 250 mg
Beilstein 7479010
Formula Weight 283.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromo-4'-(trifluoromethoxy)acetophenone
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.