Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, ≥97%, Thermo Scientific™

Catalog No. MO01244CB
Change view
Click to view available options
Amber Glass Bottle; 250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MO01244CB Amber Glass Bottle; 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01244CB Supplier Thermo Scientific Chemicals Supplier No. MO01244CB

Specifications

CAS 214360-62-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H15BBrNO2
MDL Number MFCD03412794
Quantity 250mg
Synonym 2-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-bromo-5-pyridylboronic acid pinacol ester, 2-bromopyridine-5-boronic acid pinacol ester, 2-6-bromopyridin-3-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-bromo-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-bromopyridine-3-boronic acid pinacol ester, 6-bromopyridine-3-boronicacidpinacolester, 2-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-bromopyridine-5-boronic acid pinacolate, pyridine, 2-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
InChI Key MEGBELYYUPCOIQ-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(Br)C=C1
IUPAC Name 2-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol) 283.96
PubChem CID 2794715
Formula Weight 283.96
Chemical Name or Material 2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.