Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-chloroaniline 97.0+%, TCI America™
SDP

Catalog No. B31821G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31821G 1 g
B31825G 5 g
B318225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B31821G Supplier TCI America Supplier No. B31821G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 823-57-4
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00672942
InChI Key NLEZSQHAFMZAGU-UHFFFAOYSA-N
PubChem CID 11310276
IUPAC Name 2-bromo-5-chloroaniline
SMILES C1=CC(=C(C=C1Cl)N)Br

Specifications

Melting Point 38°C
Boiling Point 128°C
Quantity 1 g
Formula Weight 206.47
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-5-chloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.