Learn More
eMolecules​ 2-Bromo-5-(difluoromethoxy)aniline | 1261875-41-5 | MFCD18390858 | 1g
Supplier:  eMolecules​ 562428182
Encompass_Preferred
Apollo Scientific | 2-Bromo-5-(difluoromethoxy)aniline | 1g | 562428182 | PC501748 | 95.000 | 1261875-41-5 | MFCD18390858 | 238.032 | C7H6BrF2NO
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-215-4029