Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-fluoro-3-nitropyridine, 95%, Thermo Scientific™

Catalog No. AAH64208MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64208MD 250 mg
AAH6420803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64208MD Supplier Thermo Scientific Chemicals Supplier No. H64208MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652160-72-0
Molecular Formula C5H2BrFN2O2
Molecular Weight (g/mol) 220.99
MDL Number MFCD05662387
InChI Key XJFDIIHIXNIWQH-UHFFFAOYSA-N
Synonym 2-bromo-3-nitro-5-fluoropyridine, 2-bromo-5-fluoro-2-nitropyrdine, 2-bromo-5-fluoro-3-nitro-pyridine, pyridine,2-bromo-5-fluoro-3-nitro, pyridine, 2-bromo-5-fluoro-3-nitro, pubchem14284, 2-bromo-3-nitro-5-fluoro pyridine, 2-bromanyl-5-fluoranyl-3-nitro-pyridine
PubChem CID 20791243
IUPAC Name 2-bromo-5-fluoro-3-nitropyridine
SMILES [O-][N+](=O)C1=CC(F)=CN=C1Br

Specifications

Color Orange
Quantity 250 mg
Beilstein 14253146
Solubility Information Slightly soluble in water.
Formula Weight 220.99
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-fluoro-3-nitropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.