Learn More
eMolecules​ 1196152-88-1 | 2-Bromo-5-fluoropyridin-4-ol | MFCD13189884 | 1g
Supplier:  eMolecules​ 525203481
Encompass_Preferred
Ambeed | 2-Chloro-1-ethynyl-4-nitrobenzene | 100mg | 624122595 | A339997 | 1260829-25-1 | MFCD11036434 | 181.580 | C8H4ClNO2
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-213-9087