Learn More
Medchemexpress LLC 2-Bromo-5-fluoropyridine | 41404-58-4 | 99.9% | 175.99 | 50 G
SDP
Catalog No. 5000403527
Encompass_Preferred
2-Bromo-5-fluoropyridine is an intermediate that can undergo Suzuki coupling reaction with Boronic acid and is utilized in the synthesis of GSK1292263A, a GPR119 receptor agonist. This product is intended for research use only.
  • Intermediate for chemical synthesis.
  • Can be used in Suzuki coupling reactions.
  • Used in the synthesis of GSK1292263A, a GPR119 receptor agonist.
Catalog No. 50-004-03527 Supplier Medchemexpress LLC Supplier No. HY3274850G
2-Bromo-5-fluoropyridine is an intermediate that can undergo Suzuki coupling reaction with Boronic acid and is utilized in the synthesis of GSK1292263A, a GPR119 receptor agonist. This product is intended for research use only.
  • Intermediate for chemical synthesis.
  • Can be used in Suzuki coupling reactions.
  • Used in the synthesis of GSK1292263A, a GPR119 receptor agonist.