Learn More
Sigma Aldrich 2-BROMO-5-HYDROXY-3-METHYLPYRIDINE
Catalog No. AGN000551G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
5 mL
Catalog No. Quantity
AGN000551G 5 mL
1 options
Catalog No. AGN000551G Supplier Sigma Aldrich Supplier No. AGN000551G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
  • 3-bromothianaphthene is a heteroaryl halide
  • It undergoes suzuki-miyaura reaction with phenylboronic acid (pba) or 3-thienylboronic acid in the presence of a novel heterogeneous pd catalyst [pd@pdeb, pdeb=poly(1,3-diethynylbenzene)]
  • The substitution reaction of 3-bromothianaphthene with piperidine to form 3-piperidinothianaphthene as the major product has been reported.3-bromothianaphthene may be used in the synthesis of (benzo[b]thiophen-3-yl)trimethylstannane
Specifications
Quantity 5 mL