Learn More
Medchemexpress LLC 2-Bromo-5-hydroxybenzaldehyde | 2973-80-0 | 201.019 | 25 G
SDP
Catalog No. 5000448137
Encompass_Preferred
2-Bromo-5-hydroxybenzaldehyde is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a useful reactant in the synthesis of PDE4 inhibitors, BCL-XL, and cancer cell growth inhibitors. It is also used in the preparation of many pharmaceutical agents.
  • Biochemical reagent for life science related research.
  • Useful reactant for synthesizing PDE4 inhibitors.
  • Useful reactant for synthesizing BCL-XL.
  • Useful reactant for synthesizing cancer cell growth inhibitors.
  • Used in the preparation of various pharmaceutical agents.
Catalog No. 50-004-48137 Supplier Medchemexpress LLC Supplier No. 25GHYW007430
2-Bromo-5-hydroxybenzaldehyde is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a useful reactant in the synthesis of PDE4 inhibitors, BCL-XL, and cancer cell growth inhibitors. It is also used in the preparation of many pharmaceutical agents.
  • Biochemical reagent for life science related research.
  • Useful reactant for synthesizing PDE4 inhibitors.
  • Useful reactant for synthesizing BCL-XL.
  • Useful reactant for synthesizing cancer cell growth inhibitors.
  • Used in the preparation of various pharmaceutical agents.