Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 2-Bromo-5-methoxybenzaldehyde, 7507-86-0, MFCD00463817, 5g

Supplier:  Matrix Scientific 0017915G

Encompass_Preferred

Molecular Formula C8H7BrO2, Purity 97%, Molecular Weight 215.05, Melting Point 73-76°

Catalog No. 50-011-3431


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.