Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-methylaniline, 97%, Thermo Scientific Chemicals

Catalog No. p-7053890
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6163906 5 g
AAH6163914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6163906 Supplier Thermo Scientific Chemicals Supplier No. H6163906

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53078-85-6
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD01806278
InChI Key QTAQWOXSUFGGKH-UHFFFAOYSA-N
Synonym 2-bromo-5-methylbenzenamine, 2-bromo-5-methylphenylamine, 2-bromo-5-methyl-phenylamine, benzenamine, 2-bromo-5-methyl, 3-amino-4-bromotoluene, 2-bromo-5-methyl-aniline, 6-bromo-m-toluidine, 2-bromo-5-methyl aniline, pubchem4337, 2-bromo-5-methylbenz
PubChem CID 6422065
IUPAC Name 2-bromo-5-methylaniline
SMILES CC1=CC(=C(C=C1)Br)N

Specifications

Density 1.486 g/mL
Flash Point >110°C (230°F)
Refractive Index 1.603
Quantity 5 g
UN Number UN2811
Formula Weight 186.05
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-methylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.