Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-nitrofuran 98.0+%, TCI America™
SDP

Catalog No. B14705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B14705G 5 g
B14701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B14705G Supplier TCI America Supplier No. B14705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 823-73-4
Molecular Formula C4H2BrNO3
Molecular Weight (g/mol) 191.97
MDL Number MFCD00142879
InChI Key QGTDMAKRJHGXOF-UHFFFAOYSA-N
PubChem CID 233582
IUPAC Name 2-bromo-5-nitrofuran
SMILES [O-][N+](=O)C1=CC=C(Br)O1

Specifications

Melting Point 47°C
Color Yellow
Boiling Point 118°C
Quantity 5 g
Formula Weight 191.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-nitrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.