Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-chloro-4-nitroaniline 98.0+%, TCI America™
SDP

Catalog No. B177125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B177125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B177125G Supplier TCI America Supplier No. B177125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99-29-6
Molecular Formula C6H4BrClN2O2
Molecular Weight (g/mol) 251.464
MDL Number MFCD00014770
InChI Key GLUCALKKMFBJEB-UHFFFAOYSA-N
Synonym 2-chloro-4-nitro-6-bromoaniline, benzenamine, 2-bromo-6-chloro-4-nitro, 6-bromo-2-chloro-4-nitroaniline, 6-bromo-2-chloro-4-nitrophenylamine, 2-bromo-4-nitro-6-chloroaniline, aniline, 2-bromo-6-chloro-4-nitro, 2-bromo-6-chloro-4-nitro-benzenamin, 2-bromo-6-chloro-4-nitro-phenylamine, pubchem2333, acmc-209sc7
PubChem CID 66832
IUPAC Name 2-bromo-6-chloro-4-nitroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Br)[N+](=O)[O-]

Specifications

Melting Point 176°C
Color Yellow
Quantity 25 g
Formula Weight 251.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-chloro-4-nitroaniline
EINECSNumber (3)-0425
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.