Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-chloropyridine 98.0+%, TCI America™
SDP

Catalog No. B36091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36091G 1 g
B36095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36091G Supplier TCI America Supplier No. B36091G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5140-72-7
Molecular Formula C5H3BrClN
Molecular Weight (g/mol) 192.44
MDL Number MFCD00181262
InChI Key JWTZSVLLPKTZJP-UHFFFAOYSA-N
Synonym pyridine, 2-bromo-6-chloro, abbypharma ap-18-5384, pubchem9626, 2-chloro-6-bromopyridine, acmc-1awk4, 2-bromo-6-chloro-pyridine, 2-bromo-6-chloropyridine?, ksc493o0h, 2-bromo-6-chloropyridine
PubChem CID 78820
IUPAC Name 2-bromo-6-chloropyridine
SMILES C1=CC(=NC(=C1)Br)Cl

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 1 g
Formula Weight 192.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.