Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-fluorobenzonitrile 97.0+%, TCI America™
SDP

Catalog No. B31835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31835G Supplier TCI America Supplier No. B31835G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79544-27-7
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD06797753
InChI Key IELGUZKHALDFOO-UHFFFAOYSA-N
Synonym 2-fluoro-6-bromobenzonitrile, 6-bromo-2-fluorobenzonitrile, 2-bromo-6-fluoro-benzonitrile, 2-cyano-3-fluoro-1-bromobenzene, benzonitrile, 2-bromo-6-fluoro, 3-bromo-2-cyanofluorobenzene, 6-bromo-2-fluorobenzenecarbonitrile, 2-cyano-3-fluorobromobenzene, pubchem2221, acmc-209pho
PubChem CID 2783139
IUPAC Name 2-bromo-6-fluorobenzonitrile
SMILES FC1=C(C#N)C(Br)=CC=C1

Specifications

Melting Point 61°C
Quantity 5 g
Formula Weight 200.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-fluorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.