Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromo-6-fluoropyridine

Catalog No. CDS002875 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS002875 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS002875 Supplier Sigma Aldrich Supplier No. CDS002875100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-chlorodiphenyl ether is halogenated diphenyl ether
  • It is an estrogenic contaminant and its effect on the growth of endometrial implants has been investigated
  • Dehalogenations of 4-chlorodiphenyl ether at 375c over cao/ca(oh)2 has been reported.4-chlorodiphenyl ether can be used in the preparation of 4α-chloro-2,2α,3,3α,4,5,5α,6,6α-nonabromodiphenyl ether (cl-bde-208), an internal standard used in the analysis of highly brominated diphenyl ethers

Specifications

CAS 144100-07-2
Molecular Formula C5H3BrFN
Linear Formula C5H3BrFN
MDL Number MFCD03095258
Quantity 1 g
Molecular Weight (g/mol) 175.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.