Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-fluorotoluene 98.0+%, TCI America™
SDP

Catalog No. B23921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23921G Supplier TCI America Supplier No. B23921G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1422-54-4
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00051438
InChI Key DJGXPFQIMLEVPA-UHFFFAOYSA-N
Synonym 2-bromo-6-fluorotoluene, 2-fluoro-6-bromotoluene, 1-bromo-3-fluoro-2-methyl-benzene, 6-bromo-2-fluorotoluene, 2-fluoro-6-bromo toluene, benzene, 1-bromo-3-fluoro-2-methyl, pubchem3236, acmc-1bwk3, 2-bromo-6-fluoro-toluene
PubChem CID 2736335
IUPAC Name 1-bromo-3-fluoro-2-methylbenzene
SMILES CC1=C(C=CC=C1Br)F

Specifications

Color Yellow
Boiling Point 180°C
Quantity 1 g
Formula Weight 189.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-6-fluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.