Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-iodopyridine, 97%

Catalog No. AAH3447806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3447806 5 g
AAH3447803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3447806 Supplier Thermo Scientific Chemicals Supplier No. H3447806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 234111-08-1 | C5H3BrIN | 283.89 g/mol

6-trifluoromethyl-2-pyridinecarboxylic acid (4; 87%) was obtained from 2-bromo-6-(trifluoromethyl)pyridine, which was in turn derived from 2-bromo-6-iodopyridine by trifluoromethylating displacement of iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-trifluoromethyl-2-pyridinecarboxylic acid (4; 87%) was obtained from 2-bromo-6-(trifluoromethyl)pyridine, which was in turn derived from 2-bromo-6-iodopyridine by trifluoromethylating displacement of iodine.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 234111-08-1
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.89
MDL Number MFCD08059557
InChI Key KJOQMWRTNVBXEV-UHFFFAOYSA-N
Synonym pyridine, 2-bromo-6-iodo, 2-bromo-6-iodo-pyridine, 6-bromo-2-iodopyridine, pyridine,2-bromo-6-iodo
PubChem CID 11312130
IUPAC Name 2-bromo-6-iodopyridine
SMILES BrC1=NC(I)=CC=C1

Specifications

Melting Point 139°C to 143°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 283.89
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-iodopyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.