Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-iodopyridine 98.0+%, TCI America™
SDP

Catalog No. B48155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48155G 5 g
B48151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48155G Supplier TCI America Supplier No. B48155G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 234111-08-1
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.89
MDL Number MFCD08059557
InChI Key KJOQMWRTNVBXEV-UHFFFAOYSA-N
Synonym pyridine, 2-bromo-6-iodo, 2-bromo-6-iodo-pyridine, 6-bromo-2-iodopyridine, pyridine,2-bromo-6-iodo
PubChem CID 11312130
IUPAC Name 2-bromo-6-iodopyridine
SMILES BrC1=NC(I)=CC=C1

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 5 g
Formula Weight 283.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.