Learn More
Medchemexpress LLC 2-bromo-6-methoxynaphthalene | 5111-65-9 | MFCD00004062 | 99.8% | 237.09 g/mol | C11H9BrO | 25g
SDP
Catalog No. 5000269380
Encompass_Preferred
2-Bromo-6-methoxynaphthalene is an active intermediate in the production of anti-inflammatory agents like Naproxen and Nabumetone by Heck reaction 2-Bromo-6-methoxynaphthalene has potential anti-inflammatory properties and Tyrosine-protein inhibitor properties 2-Bromo-6-methoxynaphthalene can be used for the research of cancer[1]
Catalog No. 50-002-69380 Supplier Medchemexpress LLC Supplier No. HYY003825G
Add to Cart
Add to Cart
2-Bromo-6-methoxynaphthalene is an active intermediate in the production of anti-inflammatory agents like Naproxen and Nabumetone by Heck reaction 2-Bromo-6-methoxynaphthalene has potential anti-inflammatory properties and Tyrosine-protein inhibitor properties 2-Bromo-6-methoxynaphthalene can be used for the research of cancer[1]