Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-pyrrolidin-1-ylpyridine, ≥97%, Thermo Scientific™

Catalog No. CC38010DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC38010DE 5 g
CC38010CB 250 mg
CC38010DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC38010DE Supplier Thermo Scientific Chemicals Supplier No. CC38010DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 230618-41-4
Molecular Formula C9H11BrN2
Molecular Weight (g/mol) 227.105
MDL Number MFCD08271892
InChI Key HZOWJDJNPISFJC-UHFFFAOYSA-N
Synonym 2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl
PubChem CID 15470277
IUPAC Name 2-bromo-6-pyrrolidin-1-ylpyridine
SMILES C1CCN(C1)C2=NC(=CC=C2)Br

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 5 g
Formula Weight 227.1
Percent Purity 90%
Chemical Name or Material 2-Bromo-6-pyrrolidin-1-ylpyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.