Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-7-iodofluorene 98.0+%, TCI America™
SDP

Catalog No. B44315G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44315G 5 g
B443125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44315G Supplier TCI America Supplier No. B44315G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123348-27-6
Molecular Formula C13H8BrI
Molecular Weight (g/mol) 371.02
MDL Number MFCD12024276
InChI Key GWUPMAMGLLLLHO-UHFFFAOYSA-N
PubChem CID 14290982
IUPAC Name 2-bromo-7-iodo-9H-fluorene
SMILES BrC1=CC2=C(C=C1)C1=C(C2)C=C(I)C=C1

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 5 g
Formula Weight 371.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-7-iodofluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.