Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-9,9-bis(2-ethylhexyl)fluorene 98.0+%, TCI America™
SDP

Catalog No. B44411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B44411G Supplier TCI America Supplier No. B44411G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 355135-07-8
Molecular Formula C29H41Br
Molecular Weight (g/mol) 469.55
MDL Number MFCD12024275
InChI Key JAWAIXIROBPNSH-UHFFFAOYNA-N
PubChem CID 11248391
IUPAC Name 2-bromo-9,9-bis(2-ethylhexyl)-9H-fluorene
SMILES CCCCC(CC)CC1(CC(CC)CCCC)C2=CC=CC=C2C2=C1C=C(Br)C=C2

Specifications

Color Yellow
Quantity 1 g
Formula Weight 469.55
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-9,9-bis(2-ethylhexyl)fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.