Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-9,9-diethylfluorene 98.0+%, TCI America™
SDP

Catalog No. B51261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51261G 1 g
B51265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51261G Supplier TCI America Supplier No. B51261G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 287493-15-6
Molecular Formula C17H17Br
Molecular Weight (g/mol) 301.23
MDL Number MFCD09909863
InChI Key HJXPGCTYMKCLTR-UHFFFAOYSA-N
PubChem CID 12123875
IUPAC Name 2-bromo-9,9-diethyl-9H-fluorene
SMILES CCC1(CC)C2=CC=CC=C2C2=C1C=C(Br)C=C2

Specifications

Melting Point 54°C
Color Yellow
Boiling Point 163°C
Quantity 1 g
Formula Weight 301.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-9,9-diethylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.