Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 2-Bromo-9,9-dimethyl-9H-fluorene, >99%, [28320-31-2], B14107 273.168 C15H13Br

Catalog No. 501374945
Encompass_Preferred

Compound: 2-Bromo-9,9-dimethyl-9H-fluorene

CAS#: 28320-31-2

MDL#: not listed

Background: A brominated, dimethylated fluorene used as a building block in materials and pharmaceutical research. Fluorene derivatives are common in organic electronic materials, and the bromine substituent provides a reactive site for cross-coupling.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-137-4945 Supplier Frontier Specialty Chemicals Supplier No. B1410725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Bromo-9,9-dimethyl-9H-fluorene

CAS#: 28320-31-2

MDL#: not listed

Background: A brominated, dimethylated fluorene used as a building block in materials and pharmaceutical research. Fluorene derivatives are common in organic electronic materials, and the bromine substituent provides a reactive site for cross-coupling.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.