Learn More
Medchemexpress LLC 2-bromo-cyclohexane-1,3-dione | 60060-44-8 | MFCD00971951 | 99.8% | 191.02 g/mol | C6H7BrO2 | 5 G
SDP
Catalog No. 5000301689
Encompass_Preferred
2-Bromo-cyclohexane-1,3-dione is a brominated cyclohexane diketone used primarily as a synthetic intermediate in medicinal chemistry. It is suitable for small-scale synthesis and method development where a reactive brominated diketone building block is required.
  • Used as a drug intermediate in organic synthesis.
  • Brominated diketone reactive at carbonyl positions.
  • High reported purity for reliable experimental results.
  • Available in multiple small pack sizes for research use.
  • Molecular formula C6H7BrO2 and molecular weight 191.02 g/mol.
Catalog No. 50-003-01689 Supplier Medchemexpress LLC Supplier No. HYW0276905G
2-Bromo-cyclohexane-1,3-dione is a brominated cyclohexane diketone used primarily as a synthetic intermediate in medicinal chemistry. It is suitable for small-scale synthesis and method development where a reactive brominated diketone building block is required.
  • Used as a drug intermediate in organic synthesis.
  • Brominated diketone reactive at carbonyl positions.
  • High reported purity for reliable experimental results.
  • Available in multiple small pack sizes for research use.
  • Molecular formula C6H7BrO2 and molecular weight 191.02 g/mol.