Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromo-isonicotinamide

Catalog No. ADE0002371G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ADE0002371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ADE0002371G Supplier Sigma Aldrich Supplier No. ADE0002371G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The following building block was synthesized via a c-h activation method developed by yu and coworkers
  • The method was incorporated using the yu-wasa auxiliary (aldrich 791806) and i2 as the sole oxidant for ortho-iodination from the corresponding benzoic acid derivative.iodination method eases route to radiolabeled drugs

Specifications

CAS 29840-73-1
Molecular Formula C6H5BrN2O
Linear Formula C6H5BrN2O
MDL Number MFCD00234156
Quantity 1 g
Molecular Weight (g/mol) 201.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.