Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-bromo-LSD | 478-84-2 | 97.3% | 10 MG
SDP

Catalog No. 5000231519
Encompass_Preferred

2-Bromo-LSD is a lysergamide research chemical and dopamine receptor antagonist supplied as a purified powder for use as an analytical standard in pharmacological studies. It is intended for research use only and not for human or veterinary administration.

  • Analytical research standard for dopaminergic studies.
  • Reported purity 97.3% (per product data sheet).
  • Solid powder form suitable for in vitro and in vivo assays.
  • Recommended storage: protect from light and store at -20°C or colder.
  • CAS number 478-84-2 and synonym BOL-148.

Catalog No. 50-002-31519 Supplier Medchemexpress LLC Supplier No. HY12167510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Bromo-LSD is a lysergamide research chemical and dopamine receptor antagonist supplied as a purified powder for use as an analytical standard in pharmacological studies. It is intended for research use only and not for human or veterinary administration.

  • Analytical research standard for dopaminergic studies.
  • Reported purity 97.3% (per product data sheet).
  • Solid powder form suitable for in vitro and in vivo assays.
  • Recommended storage: protect from light and store at -20°C or colder.
  • CAS number 478-84-2 and synonym BOL-148.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.