Learn More
Matrix Scientific 2-Bromo-m-xylene, 576-22-7, MFCD00000075, 25g
Supplier: Matrix Scientific 05693425G
Molecular Formula C8H9Br, Purity 98%, Molecular Weight 185.07, Boiling Point 206
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.