Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-N-(3,4-dimethoxyphenyl)benzenesulfonamide, 97%, Thermo Scientific™

Catalog No. AAH60073MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60073MD 250 mg
AAH6007303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60073MD Supplier Thermo Scientific Chemicals Supplier No. H60073MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1179183-69-7
Molecular Formula C14H14BrNO4S
Molecular Weight (g/mol) 372.233
MDL Number MFCD15526207
InChI Key WPZXLTFEOQPBPJ-UHFFFAOYSA-N
Synonym 2-bromo-n-3,4-dimethoxyphenyl benzenesulfonamide, 2-bromo-n-3,4-dimethoxy-phenyl-benzenesulfonamid
PubChem CID 3872247
IUPAC Name 2-bromo-N-(3,4-dimethoxyphenyl)benzenesulfonamide
SMILES COC1=C(C=C(C=C1)NS(=O)(=O)C2=CC=CC=C2Br)OC

Specifications

Quantity 250 mg
Formula Weight 372.23
Percent Purity 97%
Chemical Name or Material 2-Bromo-N-(3,4-dimethoxyphenyl)benzenesulfonamide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.