Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-bromo-N-[(3S)-tetrahydro-2-oxo-3-furanyl]benzeneacetamide | 942296-18-6 | >98.0% | 298.13 g·mol⁻¹ | C12H12BrNO3 | 10 MG
SDP

Catalog No. 5000289913
Encompass_Preferred

AHL modulator-1 is a small-molecule research reagent that modulates N-acylated L-homoserine lactone (AHL)-mediated bacterial signalling. It is supplied as a high-purity solid for laboratory research and is not for human or animal therapeutic or diagnostic use.

  • Modulates AHL-mediated bacterial signalling
  • High purity (>98.0%)
  • Molecular formula: C12H12BrNO3
  • Molecular weight: 298.13 g·mol⁻¹
  • Supplied as a 10 mg solid for research use only

Catalog No. 50-002-89913 Supplier Medchemexpress LLC Supplier No. HY16412810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AHL modulator-1 is a small-molecule research reagent that modulates N-acylated L-homoserine lactone (AHL)-mediated bacterial signalling. It is supplied as a high-purity solid for laboratory research and is not for human or animal therapeutic or diagnostic use.

  • Modulates AHL-mediated bacterial signalling
  • High purity (>98.0%)
  • Molecular formula: C12H12BrNO3
  • Molecular weight: 298.13 g·mol⁻¹
  • Supplied as a 10 mg solid for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.