Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-n-octanoic Acid 97.0+%, TCI America™
SDP

Catalog No. B062725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B062725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B062725G Supplier TCI America Supplier No. B062725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2623-82-7
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00004219
InChI Key GTGTXZRPJHDASG-ZETCQYMHSA-N
PubChem CID 552003
IUPAC Name (2S)-2-bromooctanoic acid
SMILES CCCCCC[C@H](Br)C(O)=O

Specifications

Color Yellow
Boiling Point 164°C
Quantity 25 g
UN Number 3265
Formula Weight 223.11
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Bromo-n-octanoic Acid
EINECSNumber (2)-3392
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.