Learn More
Apexbio Technology LLC 2-BROMOACETAMIDE-1G
SDP
Catalog No. 5000568636
Encompass_Preferred
2-Bromoacetamide (CAS No 683-57-8) is utilized primarily in biomedical research as an electrophilic reagent Originating from chemical synthesis this compound acts by alkylating thiol groups in proteins and enzymes interfering with their function Often studied in the context of enzyme inhibition pathways its activity is generally observed in the nanomolar to low micromolar range In vitro it is used to investigate the modification of cysteine residues in proteins while typical applications include diverse cell models and animal studies aimed at understanding biochemical pathways and potential therapeutic targets 2-Bromoacetamide is also employed in drug screening assays to evaluate the effects of protein modification Experimental concentrations typically depend on the specific research design ensuring applicability across various experimental setups
Catalog No. 50-005-68636 Supplier Apexbio Technology LLC Supplier No. C66131000
2-Bromoacetamide (CAS No 683-57-8) is utilized primarily in biomedical research as an electrophilic reagent Originating from chemical synthesis this compound acts by alkylating thiol groups in proteins and enzymes interfering with their function Often studied in the context of enzyme inhibition pathways its activity is generally observed in the nanomolar to low micromolar range In vitro it is used to investigate the modification of cysteine residues in proteins while typical applications include diverse cell models and animal studies aimed at understanding biochemical pathways and potential therapeutic targets 2-Bromoacetamide is also employed in drug screening assays to evaluate the effects of protein modification Experimental concentrations typically depend on the specific research design ensuring applicability across various experimental setups