Learn More
Medchemexpress LLC 2-Bromoacetamide | 683-57-8 | 137.96 | 100 G
SDP
Catalog No. 5000460355
Encompass_Preferred
2-Bromoacetamide is a chemical compound primarily known as a disinfection byproduct. It is used in research for its ability to inactivate liver alcohol dehydrogenase and interfere with microtubule and actin cytoskeletal function. Note that it has also been identified as a potent developmental toxicant in animals.
  • Inactivates liver alcohol dehydrogenase
  • Interferes with microtubule and actin cytoskeletal function
  • Suitable for research applications
Catalog No. 50-004-60355 Supplier Medchemexpress LLC Supplier No. HYW007330100G
2-Bromoacetamide is a chemical compound primarily known as a disinfection byproduct. It is used in research for its ability to inactivate liver alcohol dehydrogenase and interfere with microtubule and actin cytoskeletal function. Note that it has also been identified as a potent developmental toxicant in animals.
  • Inactivates liver alcohol dehydrogenase
  • Interferes with microtubule and actin cytoskeletal function
  • Suitable for research applications