Learn More
Medchemexpress LLC 2-Bromoacetamide | 683-57-8 | 137.96 | 25 G
SDP
Catalog No. 5000448070
Encompass_Preferred
2-Bromoacetamide is a disinfection byproduct known to inactivate liver alcohol dehydrogenase and interfere with microtubule and actin cytoskeletal function. It is also a potent developmental toxicant in animals.
  • Inactivates liver alcohol dehydrogenase.
  • Reduces egg cylinder formation rate and disrupts cell lineage differentiation in mouse embryos.
  • Reduces first polar body extrusion rate and disrupts spindle structure in mouse and human oocytes.
  • Interferes with microtubule and actin cytoskeleton functions.
Catalog No. 50-004-48070 Supplier Medchemexpress LLC Supplier No. 25GHYW007330
2-Bromoacetamide is a disinfection byproduct known to inactivate liver alcohol dehydrogenase and interfere with microtubule and actin cytoskeletal function. It is also a potent developmental toxicant in animals.
  • Inactivates liver alcohol dehydrogenase.
  • Reduces egg cylinder formation rate and disrupts cell lineage differentiation in mouse embryos.
  • Reduces first polar body extrusion rate and disrupts spindle structure in mouse and human oocytes.
  • Interferes with microtubule and actin cytoskeleton functions.