Learn More
Medchemexpress LLC 2-Bromoacetamide | 683-57-8 | 8025 | 137.96 | 1 ML
SDP
Catalog No. 5000460291
Encompass_Preferred
2-Bromoacetamide is a disinfection byproduct that can inactivate liver alcohol dehydrogenase and interfere with microtubule and actin cytoskeletal function. It is also a potent developmental toxicant in animals.
  • Inactivates liver alcohol dehydrogenase
  • Reduces egg cylinder formation rate and disrupts cell lineage differentiation in mouse embryos
  • Reduces the first polar body extrusion rate, disrupts spindle structure, and interferes with microtubule and actin cytoskeleton functions in mouse and human oocytes
Catalog No. 50-004-60291 Supplier Medchemexpress LLC Supplier No. HYW00733010MM/1ML
2-Bromoacetamide is a disinfection byproduct that can inactivate liver alcohol dehydrogenase and interfere with microtubule and actin cytoskeletal function. It is also a potent developmental toxicant in animals.
  • Inactivates liver alcohol dehydrogenase
  • Reduces egg cylinder formation rate and disrupts cell lineage differentiation in mouse embryos
  • Reduces the first polar body extrusion rate, disrupts spindle structure, and interferes with microtubule and actin cytoskeleton functions in mouse and human oocytes