Learn More
Medchemexpress LLC 2-Bromoacetamide | 683-57-8 | 8025 | 137.96 | 50 G
SDP
Catalog No. 5000460356
Encompass_Preferred
2-Bromoacetamide is a disinfection byproduct known to inactivate liver alcohol dehydrogenase and interfere with microtubule and actin cytoskeletal function. It has also been identified as a potent developmental toxicant in animals.
  • Inactivates liver alcohol dehydrogenase
  • Interferes with microtubule and actin cytoskeletal function
  • Functions as a potent developmental toxicant in animals
  • Reduces egg cylinder formation rate in mouse embryos (in vitro)
  • Disrupts cell lineage differentiation in mouse embryos (in vitro)
  • Reduces first polar body extrusion rate in mouse and human oocytes (in vitro)
  • Disrupts spindle structure in mouse and human oocytes (in vitro)
Catalog No. 50-004-60356 Supplier Medchemexpress LLC Supplier No. HYW00733050G
2-Bromoacetamide is a disinfection byproduct known to inactivate liver alcohol dehydrogenase and interfere with microtubule and actin cytoskeletal function. It has also been identified as a potent developmental toxicant in animals.
  • Inactivates liver alcohol dehydrogenase
  • Interferes with microtubule and actin cytoskeletal function
  • Functions as a potent developmental toxicant in animals
  • Reduces egg cylinder formation rate in mouse embryos (in vitro)
  • Disrupts cell lineage differentiation in mouse embryos (in vitro)
  • Reduces first polar body extrusion rate in mouse and human oocytes (in vitro)
  • Disrupts spindle structure in mouse and human oocytes (in vitro)