Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Bromoacetophenone 98.0+%, TCI America™
SDP

Catalog No. B053825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B053825G 25 g
B05385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B053825G Supplier TCI America Supplier No. B053825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2142-69-0
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000067
InChI Key PIMNFNXBTGPCIL-UHFFFAOYSA-N
Synonym 2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
PubChem CID 75060
IUPAC Name 1-(2-bromophenyl)ethan-1-one
SMILES CC(=O)C1=CC=CC=C1Br

Specifications

Color Yellow
Boiling Point 136°C
Quantity 25 g
Formula Weight 199.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Bromoacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.