Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromobenzaldehyde

Catalog No. B5700125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5700125G 25 g
B5700110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5700125G Supplier Sigma Aldrich Supplier No. B5700125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-Bromobenzaldehyde

Specifications

CAS 6630-33-7
Melting Point 16°C to 19°C (lit.)
Density 1.585 g/mL (at 25°C (literature))
Boiling Point 230°C (lit.)
Molecular Formula C7H5BrO
Refractive Index n20/D 1.595 (literature)
Linear Formula BrC6H4CHO
MDL Number MFCD00003300
Quantity 25 g
Synonym 2-Formylbromobenzene
Molecular Weight (g/mol) 185.02
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.