Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzaldoxime 98.0+%, TCI America™
SDP

Catalog No. B31405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31405G Supplier TCI America Supplier No. B31405G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34158-72-0
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.04
MDL Number MFCD00082734
InChI Key PSIRFUPZHPEKAE-UHFFFAOYSA-N
Synonym 2-bromobenzaldoxime, e-2-bromobenzaldehyde oxime, 2-bromobenzaldehyde oxime, e-n-2-bromophenyl methylidene hydroxylamine, e-1-2-bromophenyl-n-hydroxymethanimine, z-n-2-bromophenyl methylidene hydroxylamine, 2-bromobenzal-doxime, pubchem3731, 2-bromophenyl hydroxyimino methane, ne-n-2-bromophenyl methylidene hydroxylamine
PubChem CID 6876022
IUPAC Name N-[(2-bromophenyl)methylidene]hydroxylamine
SMILES ON=CC1=CC=CC=C1Br

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 5 g
Formula Weight 200.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromobenzaldoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.