Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzenesulfonamide, 99%, Thermo Scientific™

Catalog No. AAL1757803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1757803 1 g
AAL1757806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1757803 Supplier Thermo Scientific Chemicals Supplier No. L1757803

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92748-09-9
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.08
MDL Number MFCD00173662
InChI Key YSFGGXNLZUSHHS-UHFFFAOYSA-N
Synonym 2-bromobenzene-1-sulfonamide, 2-bromobenzenesulphonamide, 2-bromo-benzenesulfonamide, 2-bromobenzene sulfonamide, 2-bromo benzene sulfon amide, buttpark 89\07-69, 2-bromobenzene-1-sulphonamide, bromobenzenesulfonamide, bromobenzene sulfonamide, pubchem13521
PubChem CID 2801263
IUPAC Name 2-bromobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=CC=C1Br

Specifications

Melting Point 191°C to 192°C
Quantity 1 g
Beilstein 3253041
Formula Weight 236.09
Percent Purity 99%
Chemical Name or Material 2-Bromobenzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.