Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzo[b]thiophene, ≥97%, Thermo Scientific™

Catalog No. CC12210CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC12210CB 250 mg
CC12210DA 1 g
CC12210DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC12210CB Supplier Thermo Scientific Chemicals Supplier No. CC12210CB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5394-13-8
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.092
MDL Number MFCD08435846
InChI Key WIFMYMXKTAVDSQ-UHFFFAOYSA-N
Synonym 2-bromobenzo b thiophene, benzo b thiophene, 2-bromo, 2-bromobenzothiophene, bromobenzothiophene, 2-bromobenzthiophene, bromobenzo b thiophene, 2-bromo-benzo b thiophene, 2-bromobenzo b-thiophene
PubChem CID 94786
IUPAC Name 2-bromo-1-benzothiophene
SMILES C1=CC=C2C(=C1)C=C(S2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 213.1
Percent Purity 97%
Chemical Name or Material 2-Bromobenzo[b]thiophene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.