Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobornane, technical

Catalog No. AC460740250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460740250 25 g
AC460740050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460740250 Supplier Thermo Scientific Chemicals Supplier No. 460740250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 4443-48-5 | C10H17Br | 217.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4443-48-5
Molecular Formula C10H17Br
Molecular Weight (g/mol) 217.15
InChI Key OTOQMOVZIUGCQE-UHFFFAOYSA-N
Synonym bornyl bromide, 2-bromobornane, bornyl bromide, pract., 2-bromo-1,7,7-trimethylbicyclo 2.2.1 heptane, bornylbromid, bornylbromide, bornane, 2-bromo, 3-bromo-4,7,7-trimethylbicyclo 2.2.1 heptane, 2-bromo-1,7,7-trimethylbicyclo 2.2.1 heptane #
PubChem CID 138249
IUPAC Name 3-bromo-4,7,7-trimethylbicyclo[2.2.1]heptane
SMILES CC1(C2CCC1(C(C2)Br)C)C

Specifications

Boiling Point 94.0°C to 96.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 217.15
Percent Purity ≥78.0% (GC)
Grade Technical
Chemical Name or Material 2-Bromobornane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.