Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Bromobutane

Catalog No. B59500500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B59500500G 500 g
B59500100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B59500500G Supplier Sigma Aldrich Supplier No. B59500500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Viscosities of binary mixtures of 2-bromobutane and the isomeric forms of butanol have been evaluated at 298.15 and 313.15k
  • 2,2-dibromobutane, meso-2,3-dibromobutane and dl-2,3-dibromobutane are formed as major photobromination reaction products, during its reaction with molecular bromine
  • Smaller yields of 1,2-dibromobutane and 2,2,3-tribromobutane are also obtained in this reaction.2-bromobutane may be used in the synthesis of series of non-nucleosides

Specifications

CAS 78-76-2
Density 1.255 g/mL (at 25°C (literature))
Boiling Point 91°C (lit.)
Molecular Formula C4H9Br
Refractive Index n20/D 1.437 (literature)
Linear Formula CH3CH2CHBrCH3
MDL Number MFCD00000156
Quantity 500 g
Synonym sec-Butyl bromide
Molecular Weight (g/mol) 137.02
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.