Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromochlorobenzene, 98.5%

Catalog No. AC106820250
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106820250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC106820250 Supplier Thermo Scientific Chemicals Supplier No. 106820250
Add to Cart

CAS: 694-80-4 | C6H4BrCl | 191.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 694-80-4
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.45
MDL Number MFCD00000532
InChI Key QBELEDRHMPMKHP-UHFFFAOYSA-N
Synonym 2-bromochlorobenzene, o-bromochlorobenzene, 2-chlorobromobenzene, o-chlorobromobenzene, benzene, 1-bromo-2-chloro, 2-bromo-1-chlorobenzene, 1-chloro-2-bromobenzene, benzene, bromochloro, unii-pec7z3yx6p, 1-bromo-2-chloro-benzene
PubChem CID 12754
IUPAC Name 1-bromo-2-chlorobenzene
SMILES C1=CC=C(C(=C1)Cl)Br

Specifications

Color Colorless to Orange-Yellow
CAS Min % 98.0
CAS Max % 100.0
Density 1.6490g/mL
Quantity 25 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5805 to 1.5825
Specific Gravity 1.649
Assay Percent Range 98% min. (GC)
Linear Formula BrC6H4Cl
Beilstein 05, 209
Boiling Point 204°C
Solubility Information Solubility in water: 0.045g/L (25°C)
Formula Weight 191.45
Percent Purity 98.50%
Chemical Name or Material 2-Bromochlorobenzene, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-775-1
TSCA TSCA
Recommended Storage note: product tends to discolor on standing

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.