Learn More
Selleck Chemical LLC 2-bromoethanol-d4 E8218-5MG
SDP
Catalog No. 5000041944
Encompass_Preferred
2-bromoethanol-d4 is the deuterated derivative of 2-bromoethanol 2-bromoethanol is a cytotoxic alkylating agent
Catalog No. 50-000-41944 Supplier Selleck Chemical LLC Supplier No. E82185MG
2-bromoethanol-d4 is the deuterated derivative of 2-bromoethanol 2-bromoethanol is a cytotoxic alkylating agent